Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,3-Dibromonaphthalene

Bottle
Catalog # APOSOR302155-25G
New Product
Supplier:   Apollo Scientific
2,3-Dibromonaphthalene
Catalog # APOSOR302155-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  13214-70-5
2,3-Dibromonaphthalene

Bottle

CAS Number:  13214-70-5
Price
534,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2,3-Diiodonaphthalene
  • CAS Number:13214-70-5
  • Formula:C₁₀H₆Br₂
  • MDL Number:MFCD00021589
  • Molecular Weight:285.97 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications