2,3-Dibromonaphthalene
Bottle
Catalog # APOSOR302155-100MG
New Product
Supplier: Apollo Scientific
2,3-Dibromonaphthalene
Catalog # APOSOR302155-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 13214-70-5
Jump to:
Product Details & Documents
Synonyms: 2,3-Diiodonaphthalene
- CAS Number:13214-70-5
- Formula:C₁₀H₆Br₂
- MDL Number:MFCD00021589
- Molecular Weight:285.97 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



