Order Entry
Export
Orders LinkContactUsLinkComponent
AGI-6780

AGI-6780

Bottle
Catalog # APOSBISN0109-1G
New Product
Supplier:   Apollo Scientific
AGI-6780
Catalog # APOSBISN0109-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  1432660-47-3
AGI-6780

Bottle

CAS Number:  1432660-47-3
Price
1 290,00 €
Log in for availability
1 * 1 g
Product Details & Documents

AGI-6780 is a selective inhibitor of IDH2R140Q mutant with IC50 value of 23 nM.

Synonyms:  N-Cyclopropyl-4-(thiophen-3-yl)-3-(3-(3-(trifluoromethyl)phenyl)ureido)benzenesulphonamide
  • CAS Number:1432660-47-3
  • Formula:C₂₁H₁₈F₃N₃O₃S₂
  • MDL Number:MFCD26097285
  • Molecular Weight:481.51 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications