Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Potassium iodide
Potassium iodide
Potassium iodide
Bottle
APOSIN2918-2.5KG
New Product
:
Apollo Scientific
Potassium iodide
APOSIN2918-2.5KG
:
Apollo Scientific
New Product
:
7681-11-0
Potassium iodide
Bottle
:
7681-11-0
398,00 €
1 * 2,5 kg
-
+
:
Dipotassium diiodide, Kali iodide
:
7681-11-0
:
Kl
:
MFCD00011405
:
166 g/mol
:
1330 °C
:
681 °C
:
3.13 g/cm³ (25 °C)
:
Ambient
:
231-659-4
Pack type:
Bottle