Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Nitroethanol 98%

Bottle
Catalog # APOSOR74923-500G
Irritant
New Product
Supplier:   Apollo Scientific
2-Nitroethanol 98%
Catalog # APOSOR74923-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  625-48-9
2-Nitroethanol 98%

Bottle

CAS Number:  625-48-9
Price
1 130,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/reagent.

  • CAS Number:625-48-9
  • Formula:C₂H₅NO₃
  • MDL Number:MFCD00007405
  • Molecular Weight:91,066 g/mol
  • EINECS:210-895-1
Specifications

Specifications

  • Pack type:
    Bottle

Specifications