Order Entry
Export
Orders LinkContactUsLinkComponent
2-Hydroxypyridine

2-Hydroxypyridine

Bottle
Catalog # APOSOR11197-10KG
Toxic
New Product
Supplier:   Apollo Scientific
2-Hydroxypyridine
Catalog # APOSOR11197-10KG
Supplier:  Apollo Scientific
New Product
CAS Number:  142-08-5
2-Hydroxypyridine

Bottle

CAS Number:  142-08-5
Price
1 870,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
  • CAS Number:142-08-5
  • Formula:C₅H₅NO
  • MDL Number:MFCD00006268
  • Molecular Weight:95.10 g/mol
  • Boiling Pt:280…281 °C
  • Melting Pt:105…109 °C
  • Storage Temperature:Ambient
  • EINECS:205-520-3
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications