Order Entry
Export
Orders LinkContactUsLinkComponent
(-)-Quinine

(-)-Quinine

Bottle
Catalog # APOSOR28880-100G
Irritant
Health
New Product
Supplier:   Apollo Scientific
(-)-Quinine
Catalog # APOSOR28880-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  130-95-0
(-)-Quinine

Bottle

CAS Number:  130-95-0
Price
97,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol
  • CAS Number:130-95-0
  • Formula:C₂₀H₂₄N₂O₂
  • MDL Number:MFCD00198096
  • Molecular Weight:324.42 g/mol
  • Melting Pt:173…175 °C
  • Storage Temperature:Ambient
  • EINECS:205-003-2

Documents

Specifications

Specifications

  • Pack type:
    Bottle

Specifications