Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Benzodioxane

1,4-Benzodioxane

Bottle
Catalog # APOSOR4961-500G
Irritant
New Product
Supplier:   Apollo Scientific
1,4-Benzodioxane
Catalog # APOSOR4961-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  493-09-4
1,4-Benzodioxane

Bottle

CAS Number:  493-09-4
Price
196,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  2,3-Dihydrobenzo[b][1,4]dioxine, 2,3-Dihydro-1,4-benzodioxin, Benzo-1,4-dioxane, 1,4-Benzodioxan
  • CAS Number:493-09-4
  • Formula:C₈H₈O₂
  • MDL Number:MFCD00006821
  • Molecular Weight:136.15 g/mol
  • Boiling Pt:216 °C
  • Density:1.17 g/cm³ (25 °C)
  • Flash Pt:87 °C
  • Storage Temperature:Ambient
  • EINECS:207-775-6
Specifications

Specifications

  • Pack type:
    Bottle

Specifications