Order Entry
Export
Orders LinkContactUsLinkComponent
7-Methylindole

7-Methylindole

Bottle
Catalog # APOSOR6655-500G
Irritant
New Product
Supplier:   Apollo Scientific
7-Methylindole
Catalog # APOSOR6655-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  933-67-5
7-Methylindole

Bottle

CAS Number:  933-67-5
Price
1 090,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:933-67-5
  • Formula:C₉H₉N
  • MDL Number:MFCD00005684
  • Molecular Weight:131.18 g/mol
  • Boiling Pt:266 °C
  • Melting Pt:80…84 °C
  •  :Density: 1.0202 g/cm³ (99.6 °C)
  • Storage Temperature:Ambient
  • EINECS:213-270-1
Specifications

Specifications

  • Pack type:
    Bottle

Specifications