Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dimethoxybenzaldehyde 97%

3,5-Dimethoxybenzaldehyde 97%

Bottle
Catalog # APOSOR4981-5G
Irritant
New Product
Supplier:   Apollo Scientific
3,5-Dimethoxybenzaldehyde 97%
Catalog # APOSOR4981-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  7311-34-4
3,5-Dimethoxybenzaldehyde 97%

Bottle

CAS Number:  7311-34-4
Price
24,60 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:7311-34-4
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00003366
  • Molecular Weight:166.18 g/mol
  • Boiling Pt:115…118 °C/1mmHg
  • Melting Pt:44…48 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications