Order Entry
Export
Orders LinkContactUsLinkComponent
Palmitic acid 98%

Palmitic acid 98%

Bottle
Catalog # APOSOR54453-2.5KG
Irritant
New Product
Supplier:   Apollo Scientific
Palmitic acid 98%
Catalog # APOSOR54453-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  57-10-3
Palmitic acid 98%

Bottle

CAS Number:  57-10-3
Price
254,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Hexadecanoic acid, Hexadecylic acid
  • CAS Number:57-10-3
  • Formula:C₁₆H₃₂O₂
  • MDL Number:MFCD00002747
  • Molecular Weight:256,4268 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications