Order Entry
Export
Orders LinkContactUsLinkComponent
1,6-Naphthyridine

1,6-Naphthyridine

Bottle
Catalog # APOSOR42177-5G
New Product
Supplier:   Apollo Scientific
1,6-Naphthyridine
Catalog # APOSOR42177-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  253-72-5
1,6-Naphthyridine

Bottle

CAS Number:  253-72-5
Price
1 210,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Pyrido[4,3-b]pyridine, 1,6-Diazanaphthalene
  • CAS Number:253-72-5
  • Formula:C₈H₆N₂
  • MDL Number:MFCD00059750
  • Molecular Weight:130.15 g/mol
  • Melting Pt:35 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications