Order Entry
Export
Orders LinkContactUsLinkComponent
3,4-Dimethoxybenzonitrile

3,4-Dimethoxybenzonitrile

Bottle
Catalog # APOSOR2992-2.5KG
Irritant
New Product
Supplier:   Apollo Scientific
3,4-Dimethoxybenzonitrile
Catalog # APOSOR2992-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  2024-83-1
3,4-Dimethoxybenzonitrile

Bottle

CAS Number:  2024-83-1
Price
2 000,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Veratronitrile
  • CAS Number:2024-83-1
  • Formula:C₉H₉NO₂
  • MDL Number:MFCD00001802
  • Molecular Weight:163.18 g/mol
  • Melting Pt:65…70 °C
  • Storage Temperature:Ambient
  • EINECS:217-969-2
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications