Order Entry
Export
Orders LinkContactUsLinkComponent
Tris(4-fluorophenyl)phosphine 97%

Tris(4-fluorophenyl)phosphine 97%

Bottle
Catalog # APOSPC7886C-1G
New Product
Supplier:   Apollo Scientific
Tris(4-fluorophenyl)phosphine 97%
Catalog # APOSPC7886C-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  18437-78-0
Tris(4-fluorophenyl)phosphine 97%

Bottle

CAS Number:  18437-78-0
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:18437-78-0
  • Formula:C₁₈H₁₂F₃P
  • MDL Number:MFCD00013553
  • Molecular Weight:316.26 g/mol
  • Melting Pt:80…81 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications