Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2'-Hydroxy-4'-methoxyacetophenone
2'-Hydroxy-4'-methoxyacetophenone
2'-Hydroxy-4'-methoxyacetophenone
Bottle
APOSOR015436-500G
New Product
:
Apollo Scientific
2'-Hydroxy-4'-methoxyacetophenone
APOSOR015436-500G
:
Apollo Scientific
New Product
:
552-41-0
2'-Hydroxy-4'-methoxyacetophenone
Bottle
:
552-41-0
92,70 €
1 * 500 g
-
+
:
1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
:
552-41-0
:
C₉H₁₀O₃
:
MFCD00008730
:
166.18 g/mol
:
48…50 °C
:
113 °C
:
Refrigerator
:
209-012-2
Pack type:
Bottle