Order Entry
Export
Orders LinkContactUsLinkComponent
2'-Hydroxy-4'-methoxyacetophenone

2'-Hydroxy-4'-methoxyacetophenone

Bottle
Catalog # APOSOR015436-5G
Irritant
New Product
Supplier:   Apollo Scientific
2'-Hydroxy-4'-methoxyacetophenone
Catalog # APOSOR015436-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  552-41-0
2'-Hydroxy-4'-methoxyacetophenone

Bottle

CAS Number:  552-41-0
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
  • CAS Number:552-41-0
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00008730
  • Molecular Weight:166.18 g/mol
  • Melting Pt:48…50 °C
  • Flash Pt:113 °C
  • Storage Temperature:Refrigerator
  • EINECS:209-012-2
Specifications

Specifications

  • Pack type:
    Bottle

Specifications