Order Entry
Export
Orders LinkContactUsLinkComponent
 

Pentane-1,5-dithiol 98%

Bottle
Catalog # APOSOR42200-5G
New Product
Supplier:   Apollo Scientific
Pentane-1,5-dithiol 98%
Catalog # APOSOR42200-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  928-98-3
Pentane-1,5-dithiol 98%

Bottle

CAS Number:  928-98-3
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  1,5-Dimercaptopentane
  • CAS Number:928-98-3
  • Formula:C₅H₁₂S₂
  • MDL Number:MFCD00004908
  • Molecular Weight:136,27877 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications