Order Entry
Export
Orders LinkContactUsLinkComponent
1,2-Diphenylethane-1,2-Diol 98%

1,2-Diphenylethane-1,2-Diol 98%

Bottle
Catalog # APOSOR30505-100MG
Irritant
New Product
Supplier:   Apollo Scientific
1,2-Diphenylethane-1,2-Diol 98%
Catalog # APOSOR30505-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  655-48-1
1,2-Diphenylethane-1,2-Diol 98%

Bottle

CAS Number:  655-48-1
Price
37,70 €
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:655-48-1
  • Formula:C₁₄H₁₄O₂
  • MDL Number:MFCD00004499
  • Molecular Weight:214,25976 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications