Order Entry
Export
Orders LinkContactUsLinkComponent
3,3-Dimethylindoline

3,3-Dimethylindoline

Bottle
Catalog # APOSOR313014-100MG
Irritant
New Product
Supplier:   Apollo Scientific
3,3-Dimethylindoline
Catalog # APOSOR313014-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1914-02-9
3,3-Dimethylindoline

Bottle

CAS Number:  1914-02-9
Price
21,80 €
Log in for availability
1 * 100 mg
Product Details & Documents
  • CAS Number:1914-02-9
  • Formula:C₁₀H₁₃N
  • MDL Number:MFCD11847210
  • Molecular Weight:147.22 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications