Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,3-Dibromoadamantane
1,3-Dibromoadamantane
1,3-Dibromoadamantane
Bottle
APOSOR315789-2.5KG
New Product
:
Apollo Scientific
1,3-Dibromoadamantane
APOSOR315789-2.5KG
:
Apollo Scientific
New Product
:
876-53-9
1,3-Dibromoadamantane
Bottle
:
876-53-9
1 770,00 €
1 * 2,5 kg
-
+
:
876-53-9
:
C₁₀H₁₄Br₂
:
MFCD00077218
:
294.03 g/mol
:
Ambient
Pack type:
Bottle