Order Entry
Export
Orders LinkContactUsLinkComponent
Benzil

Benzil

Bottle
Catalog # APOSOR5451-100G
Irritant
New Product
Supplier:   Apollo Scientific
Benzil
Catalog # APOSOR5451-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  134-81-6
Benzil

Bottle

CAS Number:  134-81-6
Price
24,60 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,2-Diphenyl-1,2-ethanedione
  • CAS Number:134-81-6
  • Formula:(C₆H₅)C(O)C(O)(C₆H₅)
  • MDL Number:MFCD00003080
  • Molecular Weight:210.23 g/mol
  • Melting Pt:94…95 °C
  • Flash Pt:180 °C
  • Storage Temperature:Ambient
  • EINECS:205-157-0
Specifications

Specifications

  • Pack type:
    Bottle

Specifications