Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dimethoxypyridine 96%

2,3-Dimethoxypyridine 96%

Bottle
Catalog # APOSOR6412-5G
Irritant
New Product
Supplier:   Apollo Scientific
2,3-Dimethoxypyridine 96%
Catalog # APOSOR6412-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  52605-97-7
2,3-Dimethoxypyridine 96%

Bottle

CAS Number:  52605-97-7
Price
27,50 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:52605-97-7
  • Formula:C₇H₉NO₂
  • MDL Number:MFCD03095043
  • Molecular Weight:139.15 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications