Cable Glands
Special adaptation with the installation of a Ø 4 mm cable gland, allowing the insertion of up to eight external temperature probes CG4MM, Calibration and validation procedures
Catalog # ESPICG4MM
New Product
Supplier: RAYPA
Jump to:
Product Details & Documents
Installation of a Ø2 mm or Ø4 mm cable gland to provide access to as many as eight external temperature probes for calibration and validation procedures.
- Flexible and simple installation at any time, adapting to the user's needs without complications
- Compatible with Classic line and Top line autoclaves
- Two available options: GC2MM (up to 8 × Ø2 mm probes) and GC4MM (up to 8 × Ø4 mm probes)
Specifications
- Description:Special adaptation with the installation of a Ø 4 mm cable gland, allowing the insertion of up to eight external temperature probes CG4MM
- Seq:1
- For:Calibration and validation procedures