Orders LinkContactUsLinkComponent
EasyPlus™ Titrator Easy pH
EasyPlus™ Titrator Easy pH
Titrator EasyPlus™ Easy pH, EG11-BNC sensor pH aqueous
Catalog # METT30060041
New Product
Supplier:   Mettler - Toledo
EasyPlus™ Titrator Easy pH
Catalog # METT30060041
Supplier:  Mettler - Toledo
New Product
EasyPlus™ Titrator Easy pH
EasyPlus™ Titrator Easy pH
Titrator EasyPlus™ Easy pH, EG11-BNC sensor pH aqueous
Price
4 530,00 €
Log in for availability
1 * 1 items
Product Details & Documents
The Swiss engineered EasyPlus™ Titrator from METTLER TOLEDO offers a complete package for your routine application. Operation does not get simpler or more intuitive than this, thanks to the smartphone like iTitrate™ user interface and iTitrate™ intelligence.
  • The Easy pH Titrator simplifies and speeds up automated acid/base endpoint or inflection/equivalence point titrations
Affordable Swiss technology; Swiss engineering and the highest quality parts and materials make the compact and accurate titrator a perfect addition to any quality laboratory. Quick start with iTitrate™ operation; Operation is simple with the Apps based user interface and intuitive system menu navigation; save training time and make daily tasks simpler and quicker.
Ready for your sample with iTitrate™ intelligence; The built-in intelligence requires only a few parameters to be set before the instrument is ready for titration. Support; Take advantage of our internet based support. Videos, FAQ’s and a multitude of applications are just a click away. Its choice of aqueous and non-aqueous pH electrodes makes this titrator ideal for the following applications and many more: Acidity content of juices, wines and milk; Alkalinity of cleaning reagents and chemicals; Free fatty acid content of cooking oils; Alkalinity (m-value) and acidity (p-value) of water.
Burette volume: 20 ml
Application repeatability: 0,5% RDS
Titration type: pH; Direct, back compensated, blank compensated
Delivery information: Easy pH titrator include a sensor (EG11-BNC sensor pH aqueous), stirrer, 20 ml burette, tubing, operating instructions, installation and quick guide, declaration of conformity and test certificate.
Specifications
  • Description:
    Titrator EasyPlus™ Easy pH
  • Includes:
    EG11-BNC sensor pH aqueous
  • Connections:
    1×USB-A, 1×USB-B; (EasyDirect PC connection with license)
  • Cat. No.:
    METT30060041
  • Measuring range:
    ±2000 mV/0 - 14 pH
  • Temperature range:
    0...100 °C
  • mV resolution:
    ± 0,1 mV
  • Resolution:
    ± 0,1 °C
  • Display:
    109,2 mm, 480×272 pixel RGB
Accessories
Product Information
  • Catalog #
    Range
    For
    Availability
    Price
  • EG13-BNC sensor pH non-aqueous
    EG13-BNC sensor pH non-aqueous
    METT30043104
    0 - 2000 mV
    Easy Pro, Easy KFV, Easy Ox titrators
    Log in for availability
    1 * 1 items - 336,00 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyPlus™ burette, 10 ml
    EasyPlus™ burette, 10 ml
    METT30043900
    Log in for availability
    1 * 1 items - 400,00 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyPlus™ bottle head
    EasyPlus™ bottle head
    METT30060023
    Log in for availability
    1 * 1 items - 29,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Siphon tips (5 pcs.)
    Siphon tips (5 pcs.)
    662-1631
    Log in for availability
    1 * 5 items - 131,00 €
Product Information
  • Catalog #
    Capacity
    Graduation Interval
    Availability
    Price
  • Plastic beaker (80 ml), 120 pcs.
    Plastic beaker (80 ml), 120 pcs.
    661-0175
    100 ml
    20 ml
    Log in for availability
    1 * 120 items - 113,00 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyPlus™ titration head GT
    EasyPlus™ titration head GT
    METT30041102
    Log in for availability
    1 * 1 items - 223,00 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyPlus™ valve
    EasyPlus™ valve
    METT30042860
    Log in for availability
    1 * 1 items - 380,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Compact printer USB-P25
    Compact printer USB-P25
    661-0157
    Log in for availability
    1 * 1 items - 1 000,00 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyStir GT
    EasyStir GT
    METT30065467
    Log in for availability
    1 * 1 items - 264,00 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyPlus™ burette, 20 ml
    EasyPlus™ burette, 20 ml
    METT30043901
    Log in for availability
    1 * 1 items - 400,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Bottle adapter, Merck
    Bottle adapter, Merck
    662-1636
    Log in for availability
    1 * 1 items - 71,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Bottle adaptor, Fischer
    Bottle adaptor, Fischer
    662-1637
    Log in for availability
    1 * 1 items - 70,60 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyPlus™ insert set titration head GT
    EasyPlus™ insert set titration head GT
    METT30057635
    Log in for availability
    1 * 1 items - 29,80 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyPlus™ drying tube
    EasyPlus™ drying tube
    METT30044701
    Log in for availability
    1 * 1 items - 23,20 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyDirect instrument license for 3 titrators per PC
    EasyDirect instrument license for 3 titrators per PC
    METT30065449
    Log in for availability
    1 * 1 items - 956,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Temperature sensor 30K
    Temperature sensor 30K
    663-0009
    Log in for availability
    1 * 1 items - 128,00 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyPlus™ tubing set instrument
    EasyPlus™ tubing set instrument
    METT30065464
    Log in for availability
    1 * 1 items - 153,00 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyPlus™ burette cover set
    EasyPlus™ burette cover set
    METT30057633
    Log in for availability
    1 * 1 items - 78,30 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyPlus™ burette, 50 ml
    EasyPlus™ burette, 50 ml
    METT30043902
    Log in for availability
    1 * 1 items - 400,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Glass beakers (80 ml), 20 pcs.
    Glass beakers (80 ml), 20 pcs.
    661-0156
    Log in for availability
    1 * 20 items - 1 310,00 €
Product Information
  • Catalog #
    Range
    For
    Availability
    Price
  • EG11-BNC sensor pH aqueous
    EG11-BNC sensor pH aqueous
    METT30043103
    ±2000 mV/0 - 14 pH
    Easy Pro, Easy EPM, Easy pH titrators
    Log in for availability
    1 * 1 items - 244,00 €
Product Information
  • Catalog #
    Availability
    Price
  • EasyPump
    EasyPump
    METT30064218
    Log in for availability
    1 * 1 items - 553,00 €