Undecanoic acid
Undecanoic acid
Catalog # EHERC17896400
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCCCCCCCCCC(=O)O
- Inchi: InChI=1S/C11H22O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3,(H,12,13)
- IUPAC: undecanoic acid
Specifications
Specifications
- Description:Undecanoic acid
- Cat. No.:EHERC17896400
- Formula:C₁₁H₂₂O₂
- Storage conditions:20 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Undecanoic acid (6CI, 8CI, 9CI, ACI);1-Decanecarboxylic acid;1n-Undecoic acid;Hendecanoic acid;n-Undecanoic acid;n-Undecoic acid;n-Undecylic acid;NSC 7885;Undecylic acid
- Molecular weight:186,29
- CAS No.:112-37-8
- Shipping conditions:RT



