Tolmetin
Tolmetin
Catalog # EHERC17591600
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Cc1ccc(cc1)C(=O)c2ccc(CC(=O)O)n2C
- Inchi: InChI=1S/C15H15NO3/c1-10-3-5-11(6-4-10)15(19)13-8-7-12(16(13)2)9-14(17)18/h3-8H,9H2,1-2H3,(H,17,18)
- IUPAC: 2-[1-methyl-5-(4-methylbenzoyl)pyrrol-2-yl]acetic acid
Specifications
Specifications
- Description:Tolmetin
- Cat. No.:EHERC17591600
- Formula:C₁₅H₁₅NO₃
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08
- GHS signal:Danger
- Synonym(s):1H-Pyrrole-2-acetic acid, 1-methyl-5-(4-methylbenzoyl)-;Pyrrole-2-acetic acid, 1-methyl-5-p-toluoyl- (8CI);1-Methyl-5-(4-methylbenzoyl)-1H-pyrrole-2-acetic acid;1-Methyl-5-p-toluoylpyrrole-2-acetic acid;5-[(p-Tolyl)carbonyl]-1-methylpyrrole-2-acetic acid;McN 2559;Tolmetin;Tolmetine
- Molecular weight:257,28
- CAS No.:26171-23-3
- Shipping conditions:RT



