Tolfenamic acid
Tolfenamic acid
Catalog # EHERC17591000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Cc1c(Cl)cccc1Nc2ccccc2C(=O)O
- Inchi: InChI=1S/C14H12ClNO2/c1-9-11(15)6-4-8-12(9)16-13-7-3-2-5-10(13)14(17)18/h2-8,16H,1H3,(H,17,18)
- IUPAC: 2-(3-chloro-2-methylanilino)benzoic acid
- Packing group: III
Specifications
Specifications
- Description:Tolfenamic acid
- UN No.:2811
- Cat. No.:EHERC17591000
- Formula:C₁₄H₁₂ClNO₂
- Storage conditions:20 °C
- Hazard class description:GHS06, GHS08
- GHS signal:Danger
- Synonym(s):Tolfenamic acid;2-[(3-Chloro-2-methylphenyl)amino]benzoic acid;N-(3-Chlor-o-tolyl)-anthranilsäure
- Molecular weight:261,7
- CAS No.:13710-19-5
- Shipping conditions:RT



