Terconazole
Terconazole
Catalog # EHERC17320500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(C)N1CCN(CC1)c2ccc(OC[C@H]3CO[C@@](Cn4cncn4)(O3)c5ccc(Cl)cc5Cl)cc2
- Inchi: InChI=1S/C26H31Cl2N5O3/c1-19(2)31-9-11-32(12-10-31)21-4-6-22(7-5-21)34-14-23-15-35-26(36-23,16-33-18-29-17-30-33)24-8-3-20(27)13-25(24)28/h3-8,13,17-19,23H,9-12,14-16H2,1-2H3/t23-,26-/m0/s1
- IUPAC: 1-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-4-propan-2-ylpiperazine
Specifications
Specifications
- Description:Terconazole
- Cat. No.:EHERC17320500
- Formula:C₂₆H₃₁Cl₂N₅O₃
- Storage conditions:4 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Terconazole;1-[4-[[(2RS,4SR)-2-(2,4-Dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy]phenyl]-4-(1-methylethyl)piperazine
- Molecular weight:532,46
- CAS No.:67915-31-5
- Shipping conditions:RT



