Syringaldehyde
Syringaldehyde
Catalog # EHERC17080000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COc1cc(C=O)cc(OC)c1O
- Inchi: InChI=1S/C9H10O4/c1-12-7-3-6(5-10)4-8(13-2)9(7)11/h3-5,11H,1-2H3
- IUPAC: 4-hydroxy-3,5-dimethoxybenzaldehyde
Specifications
Specifications
- Description:Syringaldehyde
- Cat. No.:EHERC17080000
- Formula:C₉H₁₀O₄
- Storage conditions:20 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Benzaldehyde, 4-hydroxy-3,5-dimethoxy-;4-Hydroxy-3,5-dimethoxybenzaldehyde;2,6-Dimethoxy-4-formylphenol;3,5-Dimethoxy-4-hydroxybenzaldehyde;4-Formyl-2,6-dimethoxyphenol;4-Formylsyringol;Cedar aldehyde;Gallaldehyde 3,5-dimethyl ether;NSC 41153;SM 707;Syringaldehyde;Syringic aldehyde;VND 3207
- Molecular weight:182,17
- CAS No.:134-96-3
- Shipping conditions:RT



