Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sulfathiazole

Sulfathiazole
  EHERC17000200
Sulfathiazole
  EHERC17000200
 :  
Sulfathiazole
Sulfathiazole

Sulfathiazole

 :  
 
57,60 €
   
1 * 0,25 g
 
  • SMILES: Nc1ccc(cc1)S(=O)(=O)Nc2nccs2
  • Inchi: InChI=1S/C9H9N3O2S2/c10-7-1-3-8(4-2-7)16(13,14)12-9-11-5-6-15-9/h1-6H,10H2,(H,11,12)
  • IUPAC: 4-amino-N-(1,3-thiazol-2-yl)benzenesulfonamide

 

 

 

  • Description:
    Sulfathiazole
  • Cat. No.:
    EHERC17000200
  • Formula:
    C₉H₉N₃O₂S₂
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS07, GHS08
  • GHS signal:
    Danger
  • Synonym(s):
    Sulfathiazole;4-Amino-N-(thiazol-2-yl)benzenesulfonamide
  • Molecular weight:
    255,32
  • CAS No.:
    72-14-0
  • Shipping conditions:
    RT