Secnidazole
Secnidazole
Catalog # EHERC16931100
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(O)Cn1c(C)ncc1[N+](=O)[O-]
- Inchi: InChI=1S/C7H11N3O3/c1-5(11)4-9-6(2)8-3-7(9)10(12)13/h3,5,11H,4H2,1-2H3
- IUPAC: 1-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol
Specifications
Specifications
- Description:Secnidazole
- Cat. No.:EHERC16931100
- Formula:C₇H₁₁N₃O₃
- Storage conditions:4 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Secnidazole;Secnidazole
- Molecular weight:185,18
- CAS No.:3366-95-8
- Shipping conditions:RT



