Paracetamol
Paracetamol
Catalog # EHERC15846000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(=O)Nc1ccc(O)cc1
- Inchi: InChI=1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)
- IUPAC: N-(4-hydroxyphenyl)acetamide
Specifications
Specifications
- Description:Paracetamol
- Cat. No.:EHERC15846000
- Formula:C₈H₉NO₂
- Storage conditions:20 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Acetamide, N-(4-hydroxyphenyl)-;Acetanilide, 4'-hydroxy-;4'-Hydroxyacetanilide;4-(Acetylamino)phenol;4-(N-Acetylamino)phenol;4-Acetamidophenol;4-Acetaminophenol;4-Hydroxyacetanilide;APAP;Abensanil;Acamol;Acenol;Acenol (pharmaceutical);Acetagesic;Acetalgin;Acetaminofen;Acetaminophen;Algotropyl;Alpiny;Alvedon;Amadil;Anaflon;Anelix;Anhiba;Apamid;Apamide;Banesin;Ben-u-ron;Bickie-mol;Biocetamol;Calpol;Captin;Cetadol;Citramon P;Claratal;Clixodyne;Crocin;Dafalgan;Daga;Daphalgan;Datril;Dial-a-gesic;Dirox;Disprol;Doliprane;Dolprone;Duorol;Dymadon;Efferalgan;Endophy;Enelfa;Eneril;Eu-Med;Exdol;Febrilex;Febrilix;Febro-Gesic;Febrolin;Fendon;Fepanil;Finimal;Gattaphen T;Gelocatil;Hedex;Homoolan;Jin Gang;Korum;Lestemp;Liquagesic;Lonarid;Lyteca;Lyteca Syrup;Momentum;Multin;N-(4-Hydroxyphenyl)acetamide;N-Acetyl-4-aminophenol;N-Acetyl-4-hydroxyaniline;N-Acetyl-p-aminophenol;NAPA;NAPA (analgesic);NAPAP;NSC 109028;NSC 3991;Napafen;Naprinol;Nebs;Nobedon;Ortensan;Pacemo;Pacemol;Paldesic;Pamol;Panadol;Panadol Actifast;Panadol Extend;Panaleve;Panasorb;Panets;Panodil;Paracetamol;Paracetamol DC;Paracetamole;Paralen;Paramol;Paraspen;Parelan;Parmol;Pasolind N;Pedric;Perfalgan;Phenaphen;Phendon;Pinex;Pinex (pharmaceutical);Prodafalgan;Puerxitong;Pyrinazine;Resfenol;Resprin;Rhodapop NCR;Salzone;Sara;Tabalgin;Tachipirina;Tapar;Temlo;Tempanal;Tempra;Tralgon;Tylenol;Valadol;Valgesic;Vick Pyrena;p-(Acetylamino)phenol;p-Aceaminophenol;p-Acetamidophenol;p-Acetoaminophen;p-Hydroxyacetanilide;panadeine
- Molecular weight:151,16
- CAS No.:103-90-2
- Shipping conditions:RT



