n-Hexacosane
n-Hexacosane
Catalog # EHERC14191200
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCC
- Inchi: InChI=1S/C26H54/c1-3-5-7-9-11-13-15-17-19-21-23-25-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-26H2,1-2H3
- IUPAC: hexacosane
Specifications
Specifications
- Description:n-Hexacosane
- Cat. No.:EHERC14191200
- Formula:C₂₆H₅₄
- Storage conditions:20 °C
- Synonym(s):Hexacosane;NSC 122457;n-Hexacosane
- Molecular weight:366,71
- CAS No.:630-01-3
- Shipping conditions:RT



