Order Entry
Export
Orders LinkContactUsLinkComponent
Morpholine

Morpholine

Morpholine
Catalog # EHERC15330000
Supplier:   EHRENSTORFER
Morpholine
Catalog # EHERC15330000
Supplier:  EHRENSTORFER
CAS Number:  
Morpholine
Morpholine

Morpholine

CAS Number:  
Price
107,00 €
Log in for availability
1 * 1 mL
Product Details & Documents
  • SMILES: C1COCCN1
  • Inchi: InChI=1S/C4H9NO/c1-3-6-4-2-5-1/h5H,1-4H2
  • IUPAC: morpholine
  • Packing group: I
Specifications

Specifications

  • Description:
    Morpholine
  • UN No.:
    2054
  • Cat. No.:
    EHERC15330000
  • Formula:
    C₄H₉NO
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS02, GHS05, GHS06, GHS07
  • GHS signal:
    Danger
  • Synonym(s):
    Morpholine;Molsidomine Imp. E (EP);Molsidomine Impurity E
  • Molecular weight:
    87,12
  • CAS No.:
    110-91-8
  • Shipping conditions:
    RT

Specifications