Isoprene
Isoprene
Catalog # EHERC14449000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(=C)C=C
- Inchi: InChI=1S/C5H8/c1-4-5(2)3/h4H,1-2H2,3H3
- IUPAC: 2-methylbuta-1,3-diene
- Packing group: I
Specifications
Specifications
- Description:Isoprene
- UN No.:1218
- Cat. No.:EHERC14449000
- Formula:C₅H₈
- Storage conditions:–18 °C
- Hazard class description:GHS02, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):1,3-Butadiene, 2-methyl-;Isoprene (8CI);2-Methyl-1,3-butadiene;2-Methylbutadiene;3-Methyl-1,3-butadiene;Isopentadiene;NSC 9237;β-Methylbivinyl
- Molecular weight:68,12
- CAS No.:78-79-5
- Shipping conditions:Ice pack (–20 °C)



