DL-Glutamic acid
DL-Glutamic acid
Catalog # EHERC14034400
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: NC(CCC(=O)O)C(=O)O
- Inchi: InChI=1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)
- IUPAC: 2-aminopentanedioic acid
Specifications
Specifications
- Description:DL-Glutamic acid
- Cat. No.:EHERC14034400
- Formula:C₅H₉NO₄
- Storage conditions:20 °C
- Synonym(s):Glutamic acid (9CI, ACI);DL-Glutamic acid (ZCI);Glutamic acid, DL- (8CI);(±)-Glutamic acid;2-Aminopentanedioic acid;4-Amino-1,5-pentanedioic acid;DL-Glu;Glutaminic acid;NSC 206301;NSC 9967
- Molecular weight:147,13
- CAS No.:617-65-2
- Shipping conditions:RT



