Dimefox
Dimefox
Catalog # EHERC12650000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CN(C)P(=O)(F)N(C)C
- Inchi: InChI=1S/C4H12FN2OP/c1-6(2)9(5,8)7(3)4/h1-4H3
- IUPAC: N-[dimethylamino(fluoro)phosphoryl]-N-methylmethanamine
- Packing group: I
Specifications
Specifications
- Description:Dimefox
- UN No.:2810
- Cat. No.:EHERC12650000
- Formula:C₄H₁₂FN₂OP
- Storage conditions:4 °C
- Hazard class description:GHS06
- GHS signal:Danger
- Synonym(s):Phosphorodiamidic fluoride, tetramethyl- (6CI,7CI,8CI,9CI);N,N,N',N'-Tetramethylphosphorodiamidic fluoride;BFPO;Bis(dimethylamido)fluorophosphate;Bis(dimethylamido)fluorophosphine oxide;Bis(dimethylamido)phosphoryl fluoride;Bis(dimethylamino)fluorophosphine oxide;Bis(dimethylamino)phosphorofluoridate;CR 409;DIFO;Dimefox;Fluorophosphoric acid di(dimethylamide);Hanane;Pestox 14;Phosphine oxide, bis(dimethylamino)fluoro-;Terra-Sytam;Tetramethyldiamidophosphoric fluoride;Tetramethylphosphorodiamidic fluoride
- Molecular weight:154,12
- CAS No.:115-26-4
- Shipping conditions:RT



