Dibutyl sulfide
Dibutyl sulfide
EHERC12256500
Jump to:
- SMILES: CCCCSCCCC
- Inchi: InChI=1S/C8H18S/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3
- IUPAC: 1-butylsulfanylbutane
- Packing group: III
Specifications
Specifications
- Description:Dibutyl sulfide
- UN No.:3334
- Cat. No.:EHERC12256500
- Formula:C₈H₁₈S
- Storage conditions:20 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Dibutyl Sulfide;1-Butylsulfanylbutane;Dibutylsulfide
- Molecular weight:146,29
- CAS No.:544-40-1
- Shipping conditions:RT



