Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
3,3'-Thiodipropionic acid
BP0000762487
Print
3,3'-Thiodipropanoic acid
New Product
Supplier:
Apollo Scientific
3,3'-Thiodipropanoic acid
Supplier:
Apollo Scientific
New Product
Certificates
Return to Top
3,3'-Thiodipropanoic acid
Product Family Options
Jump to:
Product Family Options
About this item
3 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSOR93656-25G
Bottle
Log in
for availability
Log in
for availability
1 * 25 g
1 * 25 g - 21,80 €
21,80 €
-
+
1 * 25 g
21,80 €
-
+
Specifications:
New Product
New Product
APOSOR93656-100G
Bottle
Log in
for availability
Log in
for availability
1 * 100 g
1 * 100 g - 26,10 €
26,10 €
-
+
1 * 100 g
26,10 €
-
+
Specifications:
New Product
New Product
APOSOR93656-500G
Bottle
Log in
for availability
Log in
for availability
1 * 500 g
1 * 500 g - 75,30 €
75,30 €
-
+
1 * 500 g
75,30 €
-
+
Specifications:
New Product
New Product