Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,6-Diaminohexane ≥95%

Corrosive
Irritant
Supplier:   Apollo Scientific
1,6-Diaminohexane ≥95%
Supplier:  Apollo Scientific
Certificates
1,6-Diaminohexane ≥95%

3 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  1,6-Hexanediamine, Hexamethylenediamine
  • CAS Number:124-09-4
  • Formula:C₆H₁₆N₂
  • MDL Number:MFCD00008243
  • Molecular Weight:116,20463 g/mol
  • EINECS:204-679-6
  • UN:2280
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 21,80 €
    21,80 €
    1 * 50 g
       21,80 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 72,40 €
    72,40 €
    1 * 250 g
       72,40 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 218,00 €
    218,00 €
    1 * 1 kg
       218,00 €