Decanoic acid-methyl ester
Decanoic acid-methyl ester
Catalog # EHERC12095090
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCCCCCCCCC(=O)OC
- Inchi: InChI=1S/C11H22O2/c1-3-4-5-6-7-8-9-10-11(12)13-2/h3-10H2,1-2H3
- IUPAC: methyl decanoate
- Packing group: III
Specifications
Specifications
- Description:Decanoic acid-methyl ester
- UN No.:3082
- Cat. No.:EHERC12095090
- Formula:C₁₁H₂₂O₂
- Storage conditions:20 °C
- Hazard class description:GHS09
- GHS signal:Warning
- Synonym(s):Methyl caprate;Decanoic acid methyl ester;Methyl decanoate
- Molecular weight:186,29
- CAS No.:110-42-9
- Shipping conditions:RT



