Copper hydroxide
Copper hydroxide
Catalog # EHERC11698000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: O[Cu]O
- Inchi: InChI=1S/Cu.2H2O/h;2*1H2/q+2;;/p-2
- IUPAC: dihydroxycopper
- Packing group: II
Specifications
Specifications
- Description:Copper hydroxide
- UN No.:3288
- Cat. No.:EHERC11698000
- Formula:CuH₂O₂
- Storage conditions:20 °C
- Hazard class description:GHS05, GHS06, GHS07, GHS09
- GHS signal:Danger
- Molecular weight:97,56
- CAS No.:20427-59-2
- Shipping conditions:RT



