Chloroxynil
Chloroxynil
Catalog # EHERC11550000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Oc1c(Cl)cc(cc1Cl)C#N
- Inchi: InChI=1S/C7H3Cl2NO/c8-5-1-4(3-10)2-6(9)7(5)11/h1-2,11H
- IUPAC: 3,5-dichloro-4-hydroxybenzonitrile
- Packing group: III
Specifications
Specifications
- Description:Chloroxynil
- UN No.:2761
- Cat. No.:EHERC11550000
- Formula:C₇H₃Cl₂NO
- Storage conditions:20 °C
- Hazard class description:GHS06, GHS07
- GHS signal:Danger
- Molecular weight:188,01
- CAS No.:1891-95-8
- Shipping conditions:RT



