Bupirimate
Bupirimate
Catalog # EHERC10850000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCCCc1c(C)nc(NCC)nc1OS(=O)(=O)N(C)C
- Inchi: InChI=1S/C13H24N4O3S/c1-6-8-9-11-10(3)15-13(14-7-2)16-12(11)20-21(18,19)17(4)5/h6-9H2,1-5H3,(H,14,15,16)
- IUPAC: [5-butyl-2-(ethylamino)-6-methylpyrimidin-4-yl] N,N-dimethylsulfamate
- Packing group: III
Specifications
Specifications
- Description:Bupirimate
- UN No.:2588
- Cat. No.:EHERC10850000
- Formula:C₁₃H₂₄N₄O₃S
- Storage conditions:–18 °C
- Hazard class description:GHS06, GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Sulfamic acid, dimethyl-, 5-butyl-2-(ethylamino)-6-methyl-4-pyrimidinyl ester (9CI);Bupirimate;Buprimate;Nimrod;PP 588
- Molecular weight:316,42
- CAS No.:41483-43-6
- Shipping conditions:RT



