beta-Sitosterol
beta-Sitosterol
Catalog # EHERC16970850
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C
- Inchi: InChI=1S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30H,7-9,11-18H2,1-6H3/t20-,21-,23+,24+,25-,26+,27+,28+,29-/m1/s1
- IUPAC: (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
Specifications
Specifications
- Description:beta-Sitosterol
- Cat. No.:EHERC16970850
- Formula:C₂₉H₅₀O
- Storage conditions:4 °C
- Synonym(s):Nimbosterol;Stigmast-5-en-3beta-ol;(-)-beta-Sitosterol;(24R)-Ethylcholest-5-en-3beta-ol;(24R)-Stigmast-5-en-3beta-ol;22,23-Dihydrostigmasterol;24alpha-Ethylcholesterol;Angelicin;Azuprostat;Betaprost;Cinchol;Cupreol;Harzol;Inmunicin MAYMO;NSC 18173;NSC 49083;NSC 8096;Prostasal;Quebrachol;Rhammol;Rhamnol;SKF 14463;Sito-Lande;Sitosterol;Sobatum;Stigmasta-5-en-3beta-ol;Stigmasterol, 22,23-dihydro-;delta5-Stigmasten-3beta-ol;alpha-Dihydrofucosterol;alpha-Phytosterol;beta-Sitosterin;beta-Sitosterol
- Molecular weight:414,71
- CAS No.:83-46-5
- Shipping conditions:RT



