Order Entry
Export
Orders LinkContactUsLinkComponent
Basic Violet 3 D6

Basic Violet 3 D6

Basic Violet 3 D6
Catalog # EHERC10427505
Supplier:   EHRENSTORFER
Basic Violet 3 D6
Catalog # EHERC10427505
Supplier:  EHRENSTORFER
CAS Number:  
Basic Violet 3 D6
Basic Violet 3 D6

Basic Violet 3 D6

CAS Number:  
Price
367,00 €
Log in for availability
1 * 10 mg
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • SMILES: [Cl-].[2H]C([2H])([2H])N(c1ccc(cc1)C(=C2C=CC(=[N+](C)C)C=C2)c3ccc(cc3)N(C)C)C([2H])([2H])[2H]
  • Inchi: InChI=1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1/i1D3,2D3;
  • IUPAC: [4-[[4-[bis(trideuteriomethyl)amino]phenyl]-[4-(dimethylamino)phenyl]methylene]cyclohexa-2,5-dien-1-ylidene]-dimethyl-ammonium;chloride
  • Packing group: III
Specifications

Specifications

  • Description:
    Basic Violet 3 D6
  • UN No.:
    3077
  • Cat. No.:
    EHERC10427505
  • Formula:
    C₂₅D₆H₂₄N₃·Cl
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS05, GHS07, GHS08, GHS09
  • GHS signal:
    Danger
  • Synonym(s):
    Basic Violet 3 D6;Methan-d3-aminium, N-[4-[bis[4-(dimethylamino)phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-(methyl-d3)-, chloride (1:1)
  • Molecular weight:
    414,02
  • CAS No.:
    1266676-01-0
  • Shipping conditions:
    RT

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.