Order Entry
Export
Orders LinkContactUsLinkComponent
Basic Red 1

Basic Red 1

Basic Red 1
Catalog # EHERC10424950
Supplier:   EHRENSTORFER
Basic Red 1
Catalog # EHERC10424950
Supplier:  EHRENSTORFER
CAS Number:  
Basic Red 1
Basic Red 1

Basic Red 1

CAS Number:  
Price
98,20 €
Log in for availability
1 * 0,1 g
Product Details & Documents
  • SMILES: Cl.CCNc1cc2OC3=C\C(=N/CC)\C(=CC3=C(c4ccccc4C(=O)OCC)c2cc1C)C
  • Inchi: InChI=1S/C28H30N2O3.ClH/c1-6-29-23-15-25-21(13-17(23)4)27(19-11-9-10-12-20(19)28(31)32-8-3)22-14-18(5)24(30-7-2)16-26(22)33-25;/h9-16,29H,6-8H2,1-5H3;1H/b30-24+;
  • IUPAC: ethyl 2-[(6E)-3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydrochloride
  • Packing group: III
Specifications

Specifications

  • Description:
    Basic Red 1
  • UN No.:
    2811
  • Cat. No.:
    EHERC10424950
  • Formula:
    C₂₈H₃₀N₂O₃·ClH
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS05, GHS06, GHS07, GHS09
  • GHS signal:
    Danger
  • Synonym(s):
    Benzoic acid, o-[6-(ethylamino)-3-(ethylimino)-2,7-dimethyl-3H-xanthen-9-yl]-, ethyl ester, monohydrochloride (8CI);Rhodamine 6GCP (6CI);Xanthylium, 9-[2-(ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethyl-, chloride (9CI);6-Carboxyrhodamine;Aizen Rhodamine 6GCP;Basic Red 1;Basonyl Red 482;Basonyl Red 483;C.I. 45160;C.I. Basic Red 1;Calcozine Red 6G;Calcozine Rhodamine 6GX;Eljon Pink Toner;Exciton 590;Fanal Pink B;Fanal Pink GFK;Fanal Red 25532;Flexo Red 482;Heliostable Brilliant Pink B extra;Mitsui Rhodamine 6GCP;NSC 36345;Nyco Liquid Red GF;R 634;R 6G;Rh 6G;Rhodamin 6G;Rhodamine 4GD;Rhodamine 4GH;Rhodamine 5GDN;Rhodamine 5GDN Extra;Rhodamine 5GL;Rhodamine 6G;Rhodamine 6G Extra;Rhodamine 6G Extra Base;Rhodamine 6G chloride;Rhodamine 6GB;Rhodamine 6GBN;Rhodamine 6GD;Rhodamine 6GDN;Rhodamine 6GDN Extra;Rhodamine 6GEx ethyl ester;Rhodamine 6GH;Rhodamine 6GO;Rhodamine 6GX;Rhodamine 6JH;Rhodamine 6JH-SA;Rhodamine 6JH-SA Extra 1150;Rhodamine 6Zh-DN;Rhodamine F 5G;Rhodamine F 5GL;Rhodamine GDN;Rhodamine GDN Extra;Rhodamine Y 20-7425;Rhodamine Zh;Silosuper Pink B;Vali Fast Red 1308
  • Molecular weight:
    479,01
  • CAS No.:
    989-38-8
  • Shipping conditions:
    RT

Specifications