Atrazine
Supplier: EHRENSTORFER
About this item
- SMILES: CCNc1nc(Cl)nc(NC(C)C)n1
- Inchi: InChI=1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)
- IUPAC: 6-chloro-4-N-ethyl-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine
- Packing group: III
2 Options Available Below
Product Details & Documents
- SMILES: CCNc1nc(Cl)nc(NC(C)C)n1
- Inchi: InChI=1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)
- IUPAC: 6-chloro-4-N-ethyl-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine
- Packing group: III
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- EHERCR10330000
- EHERC10330000
- Synonym(s):
- 1,3,5-Triazine-2,4-diamine, 6-chloro-N2-ethyl-N4-(1-methylethyl)-;1,3,5-Triazine-2,4-diamine, 6-chloro-N-ethyl-N'-(1-methylethyl)- (9CI);s-Triazine, 2-chloro-4-(ethylamino)-6-(isopropylamino)- (8CI);6-Chloro-N2-ethyl-N4-(1-methylethyl)-1,3,5-triazine-2,4-diamine;1-Chloro-3-ethylamino-5-isopropylamino-2,4,6-triazine;2-Chloro-4-(ethylamino)-6-(2-propylamino)-s-triazine;2-Chloro-4-(ethylamino)-6-(isopropylamino)-s-triazine;2-Chloro-4-(ethylamino)-6-(isopropylamino)triazine;2-Chloro-4-ethylamineisopropylamine-s-triazine;2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine;2-Ethylamino-4-isopropylamino-6-chloro-1,3,5-triazine;2-Ethylamino-4-isopropylamino-6-chloro-s-triazine;6-Chloro-4-(ethylamino)-2-(isopropylamino)-s-triazine;6-Chloro-N-Ethyl-N'-(1-methylethyl)-1,3,5-triazine-2,4-diamine;A 361;AAtrex;AAtrex 4L;AAtrex Nine-O;ATZ;Aatrax;Aatrex 4LC;Aatrex L;Akticon;Aktikon;Aktikon PK;Aktinit A;Aktinit PK;Argezin;Atradex;Atragranz;Atranex;Atranex 80WP;Atraphyt;Atrasan;Atrataf;Atrazerba FL;Atrazin;Atrazine;Azoprim;Azoprim 50;CET;Chromozin;Cyazin;Farmozine;G 30027;Gesamprim;Gesaprim;Gesaprim 50;Gesaprim 500;Gesaprim L;Herbatoxol;Hungazin;Hungazin PK;NSC 163046;Nortox 500SC;Nu-Trazine;Oleogesaprim;Oleogesaprim 200;Primatol A;Primoleo;Radazin;Radazin T;Triazine A 1294;Wonuk;Zealin L;Zeapos;Zeazin;Zeazine;Zeopos
- 1,3,5-Triazine-2,4-diamine, 6-chloro-N2-ethyl-N4-(1-methylethyl)-;1,3,5-Triazine-2,4-diamine, 6-chloro-N-ethyl-N'-(1-methylethyl)- (9CI);s-Triazine, 2-chloro-4-(ethylamino)-6-(isopropylamino)- (8CI);6-Chloro-N2-ethyl-N4-(1-methylethyl)-1,3,5-triazine-2,4-diamine;1-Chloro-3-ethylamino-5-isopropylamino-2,4,6-triazine;2-Chloro-4-(ethylamino)-6-(2-propylamino)-s-triazine;2-Chloro-4-(ethylamino)-6-(isopropylamino)-s-triazine;2-Chloro-4-(ethylamino)-6-(isopropylamino)triazine;2-Chloro-4-ethylamineisopropylamine-s-triazine;2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine;2-Ethylamino-4-isopropylamino-6-chloro-1,3,5-triazine;2-Ethylamino-4-isopropylamino-6-chloro-s-triazine;6-Chloro-4-(ethylamino)-2-(isopropylamino)-s-triazine;6-Chloro-N-Ethyl-N'-(1-methylethyl)-1,3,5-triazine-2,4-diamine;A 361;AAtrex;AAtrex 4L;AAtrex Nine-O;ATZ;Aatrax;Aatrex 4LC;Aatrex L;Akticon;Aktikon;Aktikon PK;Aktinit A;Aktinit PK;Argezin;Atradex;Atragranz;Atranex;Atranex 80WP;Atraphyt;Atrasan;Atrataf;Atrazerba FL;Atrazin;Atrazine;Azoprim;Azoprim 50;CET;Chromozin;Cyazin;Farmozine;G 30027;Gesamprim;Gesaprim;Gesaprim 50;Gesaprim 500;Gesaprim L;Herbatoxol;Hungazin;Hungazin PK;NSC 163046;Nortox 500SC;Nu-Trazine;Oleogesaprim;Oleogesaprim 200;Primatol A;Primoleo;Radazin;Radazin T;Triazine A 1294;Wonuk;Zealin L;Zeapos;Zeazin;Zeazine;Zeopos
- CAS No.:
- 1912-24-9
- 1912-24-9
- Formula:
- C₈H₁₄ClN₅
- C₈H₁₄ClN₅
- Molecular weight:
- 215,68
- 215,68
- GHS signal:
- Warning
- Warning
- Hazard class description:
- GHS07, GHS08, GHS09
- GHS07, GHS08, GHS09
- UN No.:
- 3077
- 3077
- Shipping conditions:
- RT
- RT
- Storage conditions:
- 20 °C
- 20 °C
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- EHERCR10330000AtrazineSpecifications:
More
Specifications
Cat. No.EHERCR10330000Synonym(s)1,3,5-Triazine-2,4-diamine, 6-chloro-N2-ethyl-N4-(1-methylethyl)-;1,3,5-Triazine-2,4-diamine, 6-chloro-N-ethyl-N'-(1-methylethyl)- (9CI);s-Triazine, 2-chloro-4-(ethylamino)-6-(isopropylamino)- (8CI);6-Chloro-N2-ethyl-N4-(1-methylethyl)-1,3,5-triazine-2,4-diamine;1-Chloro-3-ethylamino-5-isopropylamino-2,4,6-triazine;2-Chloro-4-(ethylamino)-6-(2-propylamino)-s-triazine;2-Chloro-4-(ethylamino)-6-(isopropylamino)-s-triazine;2-Chloro-4-(ethylamino)-6-(isopropylamino)triazine;2-Chloro-4-ethylamineisopropylamine-s-triazine;2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine;2-Ethylamino-4-isopropylamino-6-chloro-1,3,5-triazine;2-Ethylamino-4-isopropylamino-6-chloro-s-triazine;6-Chloro-4-(ethylamino)-2-(isopropylamino)-s-triazine;6-Chloro-N-Ethyl-N'-(1-methylethyl)-1,3,5-triazine-2,4-diamine;A 361;AAtrex;AAtrex 4L;AAtrex Nine-O;ATZ;Aatrax;Aatrex 4LC;Aatrex L;Akticon;Aktikon;Aktikon PK;Aktinit A;Aktinit PK;Argezin;Atradex;Atragranz;Atranex;Atranex 80WP;Atraphyt;Atrasan;Atrataf;Atrazerba FL;Atrazin;Atrazine;Azoprim;Azoprim 50;CET;Chromozin;Cyazin;Farmozine;G 30027;Gesamprim;Gesaprim;Gesaprim 50;Gesaprim 500;Gesaprim L;Herbatoxol;Hungazin;Hungazin PK;NSC 163046;Nortox 500SC;Nu-Trazine;Oleogesaprim;Oleogesaprim 200;Primatol A;Primoleo;Radazin;Radazin T;Triazine A 1294;Wonuk;Zealin L;Zeapos;Zeazin;Zeazine;ZeoposCAS No.1912-24-9FormulaC₈H₁₄ClN₅Molecular weight215,68GHS signalWarningHazard class descriptionGHS07, GHS08, GHS09UN No.3077Shipping conditionsRTStorage conditions20 °C - EHERC10330000AtrazineSpecifications:
More
Specifications
Cat. No.EHERC10330000Synonym(s)1,3,5-Triazine-2,4-diamine, 6-chloro-N2-ethyl-N4-(1-methylethyl)-;1,3,5-Triazine-2,4-diamine, 6-chloro-N-ethyl-N'-(1-methylethyl)- (9CI);s-Triazine, 2-chloro-4-(ethylamino)-6-(isopropylamino)- (8CI);6-Chloro-N2-ethyl-N4-(1-methylethyl)-1,3,5-triazine-2,4-diamine;1-Chloro-3-ethylamino-5-isopropylamino-2,4,6-triazine;2-Chloro-4-(ethylamino)-6-(2-propylamino)-s-triazine;2-Chloro-4-(ethylamino)-6-(isopropylamino)-s-triazine;2-Chloro-4-(ethylamino)-6-(isopropylamino)triazine;2-Chloro-4-ethylamineisopropylamine-s-triazine;2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine;2-Ethylamino-4-isopropylamino-6-chloro-1,3,5-triazine;2-Ethylamino-4-isopropylamino-6-chloro-s-triazine;6-Chloro-4-(ethylamino)-2-(isopropylamino)-s-triazine;6-Chloro-N-Ethyl-N'-(1-methylethyl)-1,3,5-triazine-2,4-diamine;A 361;AAtrex;AAtrex 4L;AAtrex Nine-O;ATZ;Aatrax;Aatrex 4LC;Aatrex L;Akticon;Aktikon;Aktikon PK;Aktinit A;Aktinit PK;Argezin;Atradex;Atragranz;Atranex;Atranex 80WP;Atraphyt;Atrasan;Atrataf;Atrazerba FL;Atrazin;Atrazine;Azoprim;Azoprim 50;CET;Chromozin;Cyazin;Farmozine;G 30027;Gesamprim;Gesaprim;Gesaprim 50;Gesaprim 500;Gesaprim L;Herbatoxol;Hungazin;Hungazin PK;NSC 163046;Nortox 500SC;Nu-Trazine;Oleogesaprim;Oleogesaprim 200;Primatol A;Primoleo;Radazin;Radazin T;Triazine A 1294;Wonuk;Zealin L;Zeapos;Zeazin;Zeazine;ZeoposCAS No.1912-24-9FormulaC₈H₁₄ClN₅Molecular weight215,68GHS signalWarningHazard class descriptionGHS07, GHS08, GHS09UN No.3077Shipping conditionsRTStorage conditions20 °C



