2-Ethylhexylnitrate
2-Ethylhexylnitrate
Catalog # EHERC13342330
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCCCC(CC)CO[N+](=O)[O-]
- Inchi: InChI=1S/C8H17NO3/c1-3-5-6-8(4-2)7-12-9(10)11/h8H,3-7H2,1-2H3
- IUPAC: 2-ethylhexyl nitrate
- Packing group: III
Specifications
Specifications
- Description:2-Ethylhexylnitrate
- UN No.:3082
- Cat. No.:EHERC13342330
- Formula:C₈H₁₇NO₃
- Storage conditions:20 °C
- Hazard class description:GHS09
- GHS signal:Warning
- Molecular weight:175,23
- CAS No.:27247-96-7
- Shipping conditions:RT



