2,6-Dichlorophenol
2,6-Dichlorophenol
Catalog # EHERC12453000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Oc1c(Cl)cccc1Cl
- Inchi: InChI=1S/C6H4Cl2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H
- IUPAC: 2,6-dichlorophenol
- Packing group: III
Specifications
Specifications
- Description:2,6-Dichlorophenol
- UN No.:2020
- Cat. No.:EHERC12453000
- Formula:C₆H₄Cl₂O
- Storage conditions:20 °C
- Hazard class description:GHS05, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):2,6-Dichlorophenol;NSC 60647;Phloroglucinol Impurity I
- Molecular weight:163
- CAS No.:87-65-0
- Shipping conditions:RT



